Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TIA1 SIRNA Assays ›
Gene Symbol:
TIA1
Gene Name:
TIA1 cytotoxic granule-associated RNA binding protein
Gene Aliases:
TIA-1, WDM
Chromosome Location:
Chr.2: 70209444 - 70248793 on Build GRCh38
UniGene ID:
Hs.413123
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6327
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TIA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022037.2
NP_071320.2
11, 12
1219
NM_022173.2
NP_071505.2
12, 13
1252
XM_005264527.1
XP_005264584.1
11, 12
1226
XM_005264528.4
XP_005264585.1
11, 12
1164
XM_005264531.1
XP_005264588.1
13, 14
1299
XM_011533077.1
XP_011531379.1
12, 13
1259
XM_011533078.1
XP_011531380.1
11, 12
4925
XM_011533081.2
XP_011531383.1
12, 13
2025
XM_011533082.2
XP_011531384.1
10, 11
5597
XM_017004789.1
XP_016860278.1
12, 13
1081
XM_017004790.1
XP_016860279.1
11, 12
1047
XM_017004791.1
XP_016860280.1
10, 11
1131
XM_017004792.1
XP_016860281.1
11, 12
3436
XM_017004793.1
XP_016860282.1
11, 12
3261
XM_017004794.1
XP_016860283.1
11, 12
1993
XM_017004795.1
XP_016860284.1
13, 14
1259
XM_017004796.1
XP_016860285.1
11, 12
4767
XM_017004797.1
XP_016860286.1
12, 13
2563
XM_017004798.1
XP_016860287.1
12, 13
1294
XM_017004799.1
XP_016860288.1
12, 13
1256
XM_017004800.1
XP_016860289.1
11, 12
1223
GenBank mRNA
AB209211.1
-
12, 13
1393
DR157578.1
-
6, 7
591
M77142.1
-
11, 12
1058
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
fibroblast growth factor receptor signaling pathway
negative regulation of translation
negative regulation of cytokine biosynthetic process
regulation of mRNA splicing, via spliceosome
protein localization to cytoplasmic stress granule
positive regulation of epithelial cell apoptotic process
nucleotide binding
protein binding
poly(A) binding
AU-rich element binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline