Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048168.2 | NP_001041633.1 | 4, 5 | 448 | ||
NM_001277095.1 | NP_001264024.1 | 3, 4 | 407 | |||
NM_008692.4 | NP_032718.2 | 4, 5 | 521 | |||
GenBank mRNA | AB006181.1 | - | 3, 4 | 301 | ||
AK011327.1 | - | 4, 5 | 478 | |||
AK156353.1 | - | 3, 4 | 261 | |||
AK219441.1 | - | 1, 2 | 33 | |||
BC020117.1 | - | 4, 5 | 447 | 4009737 | ||
BC053723.1 | - | 4, 5 | 451 | 30053020 | ||
BC085261.1 | - | 4, 5 | 447 | 6397760 | ||
BM461330.1 | - | 2, 3 | 152 | |||
CJ092564.1 | - | 4, 5 | 498 | |||
DV650959.1 | - | 4, 5 | 460 | |||
U62297.1 | - | 3, 4 | 301 |