Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081453.1 | NP_001074922.1 | 1, 2 | 211 | ||
NM_001286079.2 | NP_001273008.1 | 2, 3 | 393 | |||
NM_001286080.2 | NP_001273009.1 | 3, 4 | 440 | |||
NM_008697.4 | NP_032723.2 | 3, 4 | 440 | |||
GenBank mRNA | AK129394.1 | - | 3, 4 | 330 | ||
AK134860.1 | - | 2, 3 | 406 | |||
AK139959.1 | - | 2, 3 | 419 | |||
AK141891.1 | - | 2, 3 | 381 | |||
AK162150.1 | - | 3, 4 | 384 | |||
AK167264.1 | - | 2, 3 | 386 | |||
AY515727.1 | - | 1, 2 | 190 | |||
BC137789.1 | - | 2, 3 | 278 | 8860808 | ||
BC144913.1 | - | 3, 4 | 325 | 9053461 | ||
BC144914.1 | - | 3, 4 | 325 | |||
CJ089455.1 | - | 2, 3 | 386 | |||
U40342.1 | - | 3, 4 | 356 |