Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301034.1 | NP_001287963.1 | 4 | 611 | ||
NM_008753.4 | NP_032779.2 | 4 | 617 | |||
GenBank mRNA | AK003452.1 | - | 4 | 589 | ||
AK010615.1 | - | 4 | 588 | |||
AK019155.1 | - | 4 | 588 | |||
AK087942.1 | - | 2 | 2002 | |||
AK146571.1 | - | 4 | 588 | |||
AK147167.1 | - | 4 | 588 | |||
AK151635.1 | - | 4 | 588 | |||
AK159849.1 | - | 4 | 588 | |||
AK159874.1 | - | 4 | 588 | |||
AK160491.1 | - | 4 | 589 | |||
AK160598.1 | - | 4 | 588 | |||
AK169201.1 | - | 4 | 588 | |||
BC002132.1 | - | 4 | 553 | |||
BC024902.1 | - | 4 | 590 | |||
BC046290.1 | - | 4 | 580 | |||
BC060035.1 | - | 4 | 555 | |||
BC091773.1 | - | 4 | 550 | |||
BC094287.1 | - | 4 | 570 | 5257966 | ||
U52822.1 | - | 4 | 519 |