Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008763.2 | NP_032789.1 | 1 | 731 | ||
GenBank mRNA | BC119298.1 | - | 1 | 731 | 8734047 | |
BC119300.1 | - | 1 | 731 | 8734049 |