Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048168.2 | NP_001041633.1 | 7 | 827 | ||
NM_001277095.1 | NP_001264024.1 | 6 | 786 | |||
NM_008692.4 | NP_032718.2 | 7 | 900 | |||
GenBank mRNA | AB006181.1 | - | 6 | 680 | ||
AK011327.1 | - | 7 | 856 | |||
AK156353.1 | - | 6 | 640 | |||
BC020117.1 | - | 7 | 826 | 4009737 | ||
BC053723.1 | - | 7 | 830 | 30053020 | ||
BC085261.1 | - | 7 | 826 | 6397760 | ||
BM461330.1 | - | 5 | 531 | |||
U62297.1 | - | 6 | 680 |