Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290680.1 | NP_001277609.1 | 5 | 1835 | ||
NM_001290681.1 | NP_001277610.1 | 5 | 1831 | |||
NM_008741.4 | NP_032767.1 | 5 | 1971 | |||
GenBank mRNA | AK011563.1 | - | 4 | 1787 | ||
AK075612.1 | - | 5 | 1928 | |||
AK088499.1 | - | 5 | 1963 | |||
BC018224.1 | - | 5 | 1903 | 4527900 | ||
U17259.1 | - | 5 | 1932 |