Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305115.1 | NP_001292044.1 | 4 | 1054 | ||
NM_001305116.1 | NP_001292045.1 | 3 | 821 | |||
NM_024025.2 | NP_076930.1 | 4 | 1057 | |||
GenBank mRNA | AB103376.1 | - | 4 | 872 | ||
AB158288.1 | - | 4 | 934 | |||
AB237597.1 | - | 4 | 869 | |||
AK055704.1 | - | 4 | 931 | |||
AY902194.1 | - | 4 | 872 | |||
BC001613.1 | - | 4 | 872 | 3535215 | ||
BC003115.1 | - | 3 | 442 | |||
BC067804.1 | - | 4 | 724 | 5262839 |