Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025074.2 | NP_001020245.1 | 8 | 1421 | ||
NM_001282961.1 | NP_001269890.1 | 8 | 1388 | |||
NM_008745.3 | NP_032771.1 | 8 | 1388 | |||
GenBank mRNA | AB377224.1 | - | 7 | 798 | ||
AB377225.1 | - | 7 | 798 | |||
AK018789.1 | - | 8 | 1319 | |||
AK028827.1 | - | 8 | 1241 | |||
AK147391.1 | - | 8 | 1219 | |||
AK147441.1 | - | 7 | 1209 | |||
AK160789.1 | - | 8 | 1389 | |||
BC052014.1 | - | 7 | 1183 | 5707891 | ||
KC855567.1 | - | 2 | 127 | |||
M33385.1 | - | 7 | 760 | |||
X17647.1 | - | 1271 |