Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042760.1 | NP_001036225.1 | 11 | 1474 | ||
NM_008767.2 | NP_032793.2 | 11 | 1434 | |||
GenBank mRNA | AB012084.1 | - | 11 | 1474 | ||
AF028739.1 | - | 11 | 1390 | |||
AF037065.1 | - | 11 | 1402 | |||
AK002460.1 | - | 11 | 1416 | |||
BC003734.1 | - | 11 | 1380 | 3599736 | ||
BC015078.1 | - | 10 | 1321 | 4224844 |