Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008777.3 | NP_032803.2 | 6 | 815 | ||
GenBank mRNA | AK145839.1 | - | 6 | 680 | ||
AK149519.1 | - | 6 | 681 | |||
BC013458.1 | - | 6 | 673 | 4223155 | ||
X51942.1 | - | 6 | 640 |