Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127714.2 | NP_001121186.1 | 8 | 8619 | ||
NM_024503.4 | NP_078779.2 | 9 | 8702 | |||
XM_011541884.2 | XP_011540186.1 | 10 | 8634 | |||
XM_017001992.1 | XP_016857481.1 | 10 | 9881 | |||
XM_017001993.1 | XP_016857482.1 | 8 | 8834 | |||
XM_017001994.1 | XP_016857483.1 | 9 | 8911 | |||
GenBank mRNA | AF278765.1 | - | 9 | 8495 |