Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008815.2 | NP_032841.2 | 2 | 273 | ||
GenBank mRNA | AK144538.1 | - | 2 | 230 | ||
AK144590.1 | - | 2 | 226 | |||
AK158497.1 | - | 1 | 292 | |||
AK166136.1 | - | 2 | 225 | |||
BY731410.1 | - | 2 | 273 |