Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008830.2 | NP_032856.2 | 3 | 280 | ||
GenBank mRNA | AK050345.1 | - | 2 | 233 | ||
AK144024.1 | - | 2 | 204 | |||
AK165484.1 | - | 2 | 204 | |||
BC150687.1 | - | 3 | 150 | 9087598 | ||
J03398.1 | - | 3 | 280 |