Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phxr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107288.1 | - | 1 | 187 | 40053033 | |
BC107289.1 | - | 1 | 187 | 40053040 | ||
BC127147.1 | - | 1 | 187 | 40053021 | ||
X12806.1 | - | 1 | 292 |