Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008850.1 | NP_032876.1 | 3 | 399 | ||
GenBank mRNA | AK144618.1 | - | 3 | 412 | ||
AK149745.1 | - | 3 | 389 | |||
AK151496.1 | - | 3 | 388 | |||
AK152089.1 | - | 3 | 412 | |||
AK152874.1 | - | 3 | 405 | |||
AK153569.1 | - | 3 | 412 | |||
AK165659.1 | - | 3 | 404 | |||
AK167760.1 | - | 3 | 412 | |||
AK168187.1 | - | 3 | 369 | |||
AK168720.1 | - | 3 | 402 | |||
AK187087.1 | - | 3 | 212 | |||
BC056171.1 | - | 3 | 354 | 5710926 | ||
S72681.1 | - | 3 | 174 | |||
U96725.1 | - | 3 | 400 |