Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008867.2 | NP_032893.1 | 2 | 357 | ||
GenBank mRNA | AK011427.2 | - | 2 | 334 | ||
AK076366.1 | - | 2 | 338 | |||
AK143853.1 | - | 2 | 336 | |||
BC048780.1 | - | 2 | 327 | |||
BC141355.1 | - | 2 | 238 | 9055967 | ||
BC141356.1 | - | 2 | 238 | 9055968 | ||
D30779.1 | - | 2 | 310 |