Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291508.1 | NP_001278437.1 | 3 | 1091 | ||
NM_001291509.1 | NP_001278438.1 | 3 | 1091 | |||
NM_008783.3 | NP_032809.1 | 3 | 1091 | |||
NM_183355.3 | NP_899198.1 | 3 | 1091 | |||
GenBank mRNA | AB221563.1 | - | 3 | 426 | ||
AF020196.1 | - | 3 | 782 | |||
AK084143.1 | - | 3 | 583 | |||
AK087587.1 | - | 2 | 214 | |||
BC002244.1 | - | 3 | 317 | 3492658 | ||
BC054376.1 | - | 3 | 1052 | |||
BC058390.1 | - | 3 | 819 | 5701148 | ||
HM369110.1 | - | 3 | 426 | |||
L27453.1 | - | 3 | 470 |