Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008808.3 | NP_032834.1 | 3, 4 | 441 | ||
GenBank mRNA | AK017169.1 | - | 1, 2 | 26 | ||
AY324648.1 | - | 3, 4 | 430 | |||
BC003817.1 | - | 3, 4 | 441 | 3495629 | ||
M29464.1 | - | 3, 4 | 321 |