Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pecam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037551.1 | - | 4 | 828 | ||
AK084527.1 | - | 3 | 417 | |||
AK169431.1 | - | 3 | 420 | |||
BC008519.1 | - | 3 | 422 | 3592726 | ||
BC085502.1 | - | 3 | 224 | 5720681 | ||
L06039.1 | - | 3 | 307 |