Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008822.2 | NP_032848.1 | 3, 4 | 430 | ||
GenBank mRNA | AK004687.1 | - | 3, 4 | 415 | ||
AK160195.1 | - | 3, 4 | 394 | |||
BC132213.1 | - | 3, 4 | 403 | 40130490 | ||
BC132503.1 | - | 3, 4 | 403 | 40130780 | ||
DV660555.1 | - | 3, 4 | 431 | |||
U69171.1 | - | 3, 4 | 421 |