Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271705.1 | NP_001258634.1 | 4 | 388 | ||
NM_008827.3 | NP_032853.1 | 4 | 721 | |||
GenBank mRNA | AK042891.1 | - | 4 | 705 | ||
AK088943.1 | - | 4 | 704 | |||
BC016567.1 | - | 4 | 693 | 2649577 | ||
X80171.1 | - | 4 | 489 | |||
X96793.1 | - | 4 | 688 |