Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 6 | 652 | ||
GenBank mRNA | AK002714.1 | - | 5 | 598 | ||
AK010619.1 | - | 6 | 595 | |||
AK150956.1 | - | 6 | 597 | |||
AK151073.1 | - | 6 | 597 | |||
AK152933.1 | - | 6 | 599 | |||
AK160490.1 | - | 6 | 591 | |||
AK168248.1 | - | 6 | 600 | |||
AK168656.1 | - | 6 | 591 | |||
BC083354.1 | - | 5 | 574 | 5715690 | ||
BC089034.1 | - | 5 | 576 | 30675988 | ||
X78682.1 | - | 5 | 580 |