Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008866.2 | NP_032892.1 | 5, 6 | 302 | ||
GenBank mRNA | AK002674.1 | - | 5, 6 | 306 | ||
AK050549.1 | - | 5, 6 | 279 | |||
AK146874.1 | - | 5, 6 | 317 | |||
AK167231.1 | - | 324 | ||||
BC013536.1 | - | 5, 6 | 302 | 4240573 | ||
BC052848.1 | - | 5, 6 | 343 | 30055025 | ||
CT010201.1 | - | 5, 6 | 281 | |||
U89352.1 | - | 5, 6 | 281 |