Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031283.2 | NP_112573.1 | 12 | 2070 | ||
XM_006712109.2 | XP_006712172.1 | 12 | 1863 | |||
GenBank mRNA | AB031046.1 | - | 12 | 1919 |