Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045558.1 | NP_001039023.1 | 13 | 1594 | ||
NM_008837.3 | NP_032863.2 | 13 | 1653 | |||
GenBank mRNA | AI893973.1 | - | 2 | 417 | ||
BC030912.1 | - | 3 | 1642 | |||
BC048788.1 | - | 13 | 1539 | 6488048 | ||
Z46720.1 | - | 12 | 1342 |