Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277898.1 | NP_001264827.1 | 10 | 1212 | ||
NM_008854.5 | NP_032880.1 | 10 | 1255 | |||
GenBank mRNA | AK217908.1 | - | 1 | 38 | ||
BC003238.1 | - | 10 | 1245 | 3497908 | ||
BC054834.1 | - | 10 | 1240 | 6439034 | ||
CB248596.1 | - | 2 | 152 | |||
CD807077.1 | - | 6 | 659 | |||
M12303.1 | - | 10 | 1232 |