Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008866.2 | NP_032892.1 | 9 | 1947 | ||
GenBank mRNA | AK002674.1 | - | 9 | 1951 | ||
AK146874.1 | - | 9 | 1962 | |||
BC013536.1 | - | 9 | 1947 | 4240573 | ||
BC052848.1 | - | 9 | 1986 | 30055025 |