Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008867.2 | NP_032893.1 | 15 | 2520 | ||
GenBank mRNA | BC141355.1 | - | 15 | 2401 | 9055967 | |
BC141356.1 | - | 15 | 2401 | 9055968 | ||
D30779.1 | - | 15 | 2473 |