Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040179.1 | NP_001035269.1 | 6 | 1304 | ||
NM_032503.2 | NP_115892.2 | 6 | 1080 | |||
GenBank mRNA | AB058849.1 | - | 5 | 989 | ||
AB060151.1 | - | 5 | 989 | |||
AF347063.1 | - | 6 | 1100 | |||
AF399937.1 | - | 5 | 989 | |||
AK123634.1 | - | 6 | 1304 | |||
AY029596.1 | - | 5 | 989 | |||
AY562946.1 | - | 5 | 989 | |||
BC095515.1 | - | 5 | 1016 | 6971912 |
Set Membership: |
G-protein coupled receptor |