Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008907.1 | NP_032933.1 | 3 | 182 | ||
GenBank mRNA | AK002236.1 | - | 3 | 185 | ||
AK007981.1 | - | 3 | 187 | |||
AK010649.1 | - | 3 | 185 | |||
AK012491.1 | - | 3 | 193 | |||
AK012531.1 | - | 3 | 185 | |||
AK012541.1 | - | 3 | 181 | |||
AK012663.1 | - | 3 | 185 | |||
AK028210.1 | - | 3 | 185 | |||
AK131942.1 | - | 3 | 185 | |||
AK136788.1 | - | 3 | 185 | |||
AK151347.1 | - | 3 | 185 | |||
AK151583.1 | - | 3 | 274 | |||
AK153030.1 | - | 3 | 184 | |||
AK160631.1 | - | 3 | 185 | |||
AK160780.1 | - | 3 | 213 | |||
AK168035.1 | - | 3 | 185 | |||
AK169813.1 | - | 3 | 185 | |||
BC013713.1 | - | 3 | 173 | |||
BC063097.1 | - | 2 | 2297 | |||
BC083076.1 | - | 3 | 173 | 6442005 | ||
BC087928.1 | - | 3 | 184 | 6533710 | ||
BC094272.1 | - | 3 | 173 | 2645114 | ||
BC094632.1 | - | 3 | 165 | 5055863 | ||
BC099478.1 | - | 3 | 176 | 30843162 | ||
DQ985731.1 | - | 3 | 141 | |||
X52803.1 | - | 3 | 182 |