Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008908.4 | NP_032934.1 | 1, 2 | 176 | ||
GenBank mRNA | AK131931.1 | - | 1, 2 | 184 | ||
AK150654.1 | - | 1, 2 | 142 | |||
AK151449.1 | - | 1, 2 | 142 | |||
AK168911.1 | - | 1, 2 | 160 | |||
BC025861.1 | - | 1, 2 | 191 | 5026674 | ||
M74227.1 | - | 1, 2 | 176 |