Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293650.1 | NP_001280579.1 | 3 | 346 | ||
NM_001293651.1 | NP_001280580.1 | 3 | 350 | |||
NM_008925.2 | NP_032951.1 | 3 | 346 | |||
GenBank mRNA | AK137661.1 | - | 3 | 313 | ||
AK153931.1 | - | 3 | 321 | |||
AK159705.1 | - | 3 | 317 | |||
AK203731.1 | - | 2 | 125 | |||
BC009816.1 | - | 3 | 279 | 4008182 | ||
BY295975.1 | - | 3 | 347 | |||
CD807078.1 | - | 3 | 315 | |||
U92794.1 | - | 3 | 299 |