Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164058.1 | NP_001157530.1 | 2 | 189 | ||
NM_008930.2 | NP_032956.1 | 2 | 186 | |||
GenBank mRNA | AF011381.1 | - | 2 | 178 | ||
AF020525.1 | - | 2 | 160 | |||
AK028371.1 | - | 2 | 187 | |||
AK141309.1 | - | 2 | 190 | |||
AK146050.1 | - | 2 | 190 | |||
BB611867.1 | - | 2 | 187 | |||
BC051671.1 | - | 2 | 171 | 30018275 |