Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008940.2 | NP_032966.1 | 2 | 590 | ||
GenBank mRNA | BC055895.1 | - | 2 | 104 | 4019947 | |
D30785.1 | - | 2 | 590 |