Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008948.2 | NP_032974.2 | 4 | 564 | ||
GenBank mRNA | AK083043.1 | - | 526 | |||
AK150189.1 | - | 4 | 500 | |||
AK151370.1 | - | 4 | 284 | |||
AK152018.1 | - | 4 | 303 | |||
AK167002.1 | - | 4 | 512 | |||
AK167061.1 | - | 4 | 509 | |||
AK168263.1 | - | 3 | 233 | |||
AK168616.1 | - | 4 | 284 | |||
BC005783.1 | - | 4 | 453 | 3603053 | ||
CX201527.1 | - | 4 | 571 | |||
D49686.1 | - | 4 | 508 |