Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012076.1 | - | 2 | 280 | ||
AK075604.1 | - | 2 | 279 | |||
AK135948.1 | - | 2 | 280 | |||
AK178461.1 | - | 3 | 311 | |||
AK178809.1 | - | 2 | 217 | |||
AK207887.1 | - | 2 | 218 | |||
AK216138.1 | - | 3 | 303 | |||
BC003793.1 | - | 2 | 247 | 3601675 | ||
U28016.1 | - | 2 | 256 |