Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008890.1 | NP_032916.1 | 3 | 852 | ||
GenBank mRNA | AK136691.1 | - | 3 | 861 | ||
BC119079.1 | - | 3 | 882 | 8733828 | ||
BC120789.1 | - | 3 | 882 | 40129712 |