Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou3f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008901.1 | NP_032927.1 | 1 | 709 | ||
GenBank mRNA | AK158302.1 | - | 1 | 742 | ||
BC052012.1 | - | 1 | 757 | |||
BC086670.1 | - | 720 | ||||
BC138657.1 | - | 1 | 784 | 8861677 | ||
BC145919.1 | - | 1 | 784 | 40131222 | ||
X66603.1 | - | 752 |