Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppargc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008904.2 | NP_032930.1 | 5 | 830 | ||
GenBank mRNA | AB061324.1 | - | 5 | 753 | ||
AF049330.1 | - | 5 | 782 | |||
AK138668.1 | - | 5 | 831 | |||
AK143753.1 | - | 5 | 782 | |||
BC066868.1 | - | 5 | 725 | 30094033 | ||
JX866948.1 | - | 5 | 718 |