Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008938.1 | NP_032964.1 | 1, 2 | 790 | ||
GenBank mRNA | AK044549.1 | - | 1, 2 | 785 | ||
AK136280.1 | - | 1, 2 | 817 | |||
BC028958.1 | - | 1, 2 | 785 | 5363367 | ||
BC085266.1 | - | 1, 2 | 789 | 6490224 | ||
X14770.1 | - | 1, 2 | 790 |