Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008981.3 | NP_033007.2 | 3, 4 | 1064 | ||
GenBank mRNA | AK053271.1 | - | 3, 4 | 1036 | ||
BC079595.1 | - | 3, 4 | 1064 | |||
L09562.1 | - | 3, 4 | 605 |