Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igdcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008988.2 | NP_033014.1 | 2 | 586 | ||
GenBank mRNA | AF026465.1 | - | 2 | 586 | ||
AK011916.1 | - | 2 | 622 | |||
AK051027.1 | - | 2 | 587 | |||
BC053057.1 | - | 2 | 563 | 5698028 | ||
BC086481.1 | - | 2 | 570 | 6505546 |