Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008996.3 | NP_033022.1 | 4 | 246 | ||
GenBank mRNA | AB232583.1 | - | 4 | 218 | ||
AF226873.1 | - | 4 | 246 | |||
AK034408.1 | - | 4 | 414 | |||
AK129477.1 | - | 4 | 404 | |||
AK152978.1 | - | 4 | 415 | |||
AK159425.1 | - | 4 | 410 | |||
AK166249.1 | - | 4 | 407 | |||
AK167150.1 | - | 4 | 414 | |||
AK197899.1 | - | 305 | ||||
BC002077.1 | - | 4 | 413 | 3592802 | ||
Y00094.1 | - | 4 | 347 |