Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008926.4 | NP_032952.3 | 17, 18 | 2304 | ||
GenBank mRNA | AK038739.1 | - | 17, 18 | 2288 | ||
AK081422.1 | - | 17, 18 | 2305 | |||
AK165904.1 | - | 2, 3 | 462 | |||
BC113205.1 | - | 17, 18 | 2215 | 40047751 | ||
L12460.1 | - | 17, 18 | 2576 |