Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSEN1 SIRNA Assays ›
Gene Symbol:
Psen1
Gene Name:
presenilin 1
Gene Aliases:
Ad3h, PS-1, PS1, S182
Chromosome Location:
Chr.12: 83688146 - 83738446 on Build GRCm38
UniGene ID:
Mm.998
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
63988
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psen1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008943.2
NP_032969.1
12
2976
Back To Top
More Information
Panther Classification:
Molecular Function -
aspartic protease
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
negative regulation of transcription from RNA polymerase II promoter
activation of MAPKK activity
blood vessel development
cell fate specification
somitogenesis
neuron migration
positive regulation of receptor recycling
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
heart looping
hematopoietic progenitor cell differentiation
T cell activation involved in immune response
myeloid leukocyte differentiation
negative regulation of protein kinase activity
protein glycosylation
proteolysis
membrane protein ectodomain proteolysis
calcium ion transport
mitochondrial transport
cellular calcium ion homeostasis
autophagy
apoptotic process
cellular response to DNA damage stimulus
response to oxidative stress
cell adhesion
negative regulation of epidermal growth factor-activated receptor activity
regulation of epidermal growth factor-activated receptor activity
Notch signaling pathway
Notch receptor processing
brain development
heart development
learning or memory
memory
embryo development
post-embryonic development
protein transport
L-glutamate transport
choline transport
synaptic vesicle targeting
single organismal cell-cell adhesion
protein processing
cerebral cortex cell migration
Cajal-Retzius cell differentiation
dorsal/ventral neural tube patterning
cerebral cortex development
neurogenesis
neuron differentiation
embryonic limb morphogenesis
forebrain development
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
endoplasmic reticulum calcium ion homeostasis
beta-amyloid formation
segmentation
intracellular signal transduction
locomotion
regulation of phosphorylation
anagen
amyloid precursor protein catabolic process
myeloid dendritic cell differentiation
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of catalytic activity
regulation of protein binding
positive regulation of MAP kinase activity
negative regulation of neuron apoptotic process
skin morphogenesis
cellular protein metabolic process
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
regulation of synaptic plasticity
thymus development
neuron development
skeletal system morphogenesis
brain morphogenesis
beta-amyloid metabolic process
epithelial cell proliferation
negative regulation of axonogenesis
positive regulation of coagulation
T cell receptor signaling pathway
neuron apoptotic process
negative regulation of ubiquitin-protein transferase activity
smooth endoplasmic reticulum calcium ion homeostasis
protein maturation
regulation of synaptic transmission, glutamatergic
canonical Wnt signaling pathway
regulation of resting membrane potential
positive regulation of dendritic spine development
calcium ion transmembrane transport
negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of apoptotic signaling pathway
endopeptidase activity
aspartic-type endopeptidase activity
calcium channel activity
protein binding
beta-catenin binding
peptidase activity
hydrolase activity
PDZ domain binding
aspartic endopeptidase activity, intramembrane cleaving
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0