Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008947.3 | NP_032973.1 | 3 | 261 | ||
GenBank mRNA | AK088411.1 | - | 4 | 291 | ||
AK133926.1 | - | 4 | 295 | |||
AK148906.1 | - | 4 | 283 | |||
AK161641.1 | - | 4 | 310 | |||
AK161723.1 | - | 4 | 311 | |||
AK166783.1 | - | 4 | 276 | |||
AK169015.1 | - | 4 | 274 | |||
BC003860.1 | - | 4 | 277 | 3486436 | ||
U39302.1 | - | 3 | 262 |