Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 2 | 522 | ||
NM_001290077.1 | NP_001277006.1 | 2 | 522 | |||
XM_005251561.1 | XP_005251618.1 | 2 | 189 | |||
XM_005251563.1 | XP_005251620.1 | 2 | 189 | |||
GenBank mRNA | AB086825.1 | - | 2 | 178 | ||
AB208796.1 | - | 2 | 200 | |||
AK291775.1 | - | 2 | 210 | |||
AK295043.1 | - | 2 | 248 | |||
BC035514.1 | - | 2 | 197 | 5228999 | ||
CA392256.1 | - | 2 | 425 | |||
DC324016.1 | - | 2 | 449 | |||
EU826591.1 | - | 2 | 72 | |||
EU826592.1 | - | 2 | 72 | |||
L06139.1 | - | 2 | 220 |