Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008976.2 | NP_033002.2 | 17, 18 | 3812 | ||
GenBank mRNA | AF170902.1 | - | 16, 17 | 2463 | ||
AF170903.1 | - | 16, 17 | 2359 | |||
AK156480.1 | - | 17, 18 | 3557 | |||
CK780322.1 | - | 2, 3 | 374 | |||
D31842.1 | - | 16, 17 | 3409 |