Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008990.3 | NP_033016.3 | 9 | 2563 | ||
GenBank mRNA | AK190540.1 | - | 1 | 269 | ||
BC046770.1 | - | 9 | 2459 | 5703547 | ||
BC059941.1 | - | 9 | 2512 | 6530953 | ||
D26107.1 | - | 9 | 2296 |